Learn More
Usbeck Laborgeraete™ Nickel-plated Brass Mohr Pinchcocks
67.50 SEK - 79.80 SEK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10569841
|
Usbeck Laborgeraete™
5531 |
50 mm |
67.50 SEK
Each |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
10018230
|
Usbeck Laborgeraete™
5534 |
80 mm |
79.80 SEK
Each |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
10078200
|
Usbeck Laborgeraete™
5532 |
60 mm |
70.70 SEK
Each |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
10560032
|
Usbeck Laborgeraete™
5533 |
70 mm |
74.75 SEK
Each |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
Specifications
Each | |
Mohr Pinchcock | |
Nickel-plated Brass |