missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX3.1 Rabbit anti-Human, Clone: 4X8T3, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | NKX3.1 |
---|---|
Clone | 4X8T3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18323735
|
Bio-Techne
NBP3-15883-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18356235
|
Bio-Techne
NBP3-15883-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKX3.1 Monoclonal antibody specifically detects NKX3.1 in Human samples. It is validated for Western BlotSpecifications
NKX3.1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
4824 | |
IgG | |
Affinity purified |
4X8T3 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-234 of human NKX3.1 (Q99801). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title