missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLGN4Y Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | NLGN4Y |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881724
|
Proteintech
13489-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871724
|
Proteintech
13489-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody can recognize both of NLGN4Y and NLGN4X at 72 kDa mature form.
Neuroligins, such as NLGN4Y, are cell adhesion molecules present at the postsynaptic side of the synapse and may be essential for the formation of functional synapses (Jamain et al., 2003 ).Specifications
NLGN4Y | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8N0W4, Q8NFZ3 | |
22829, 57502 | |
NLGN4Y Fusion Protein Ag4311 | |
Primary | |
-20°C | |
NLGN4Y |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0951, neuroligin 4, Y linked, Neuroligin Y, NLGN4Y | |
NLGN4X, NLGN4Y | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title