Learn More
DWK Life Sciences NMR brush, for cleaning NMR tubes
Stubborn soiling and deposits in NMR tubes can be removed with the NMR brush.
Brand: DWK Life Sciences 290740059
36.16 SEK valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Additional Details : Weight : 0.00200kg
Description
Nuclear magnetic resonance spectroscopy (NMR-spectroscopy) is a spectroscopic method to elucidate the structure of molecules and compounds. Due to electronic interactions of single atoms with the neighbouring atoms, structural conclusions can be drawn. In addition to structure elucidation, the concentration and dynamics of molecules can be determined with this method. This analytical method requires very precise tubes with a constant diameter, a uniform wall thickness and excellent straightness across the entire tube. Deposits and contamination in NMR tubes can be dissolved with the help of the NMR brush. Subsequent cleaning with a suitable solvent completely acidifies the NMR tube.
- Due to the accuracy of fit, the bristles rest against the inner wall of the glass and an optimal cleaning process is possible.
Specifications
Nylon | |
Curved | |
Cleaning | |
250 mm |
Stainless Steel | |
Autoclavable | |
Cleaning |