Learn More
Nonanal, 97%, Thermo Scientific Chemicals
803.51 SEK - 3230.62 SEK
Chemical Identifiers
CAS | 124-19-6 |
---|---|
Molecular Formula | C9H18O |
Molecular Weight (g/mol) | 142.242 |
MDL Number | MFCD00007030 |
InChI Key | GYHFUZHODSMOHU-UHFFFAOYSA-N |
Synonym | pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde |
PubChem CID | 31289 |
ChEBI | CHEBI:84268 |
IUPAC Name | nonanal |
SMILES | CCCCCCCCC=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11428436
|
Thermo Scientific Alfa Aesar
A15908.22 |
100 g |
803.51 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438436
|
Thermo Scientific Alfa Aesar
A15908.36 |
500 g |
3230.62 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nonanal is used for fragranced end-products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNonanal is used for fragranced end-products.
Solubility
Insoluble in water.
Notes
Incompatible with reducing agents, strong basesand strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Chemical Identifiers
124-19-6 | |
142.242 | |
GYHFUZHODSMOHU-UHFFFAOYSA-N | |
31289 | |
nonanal |
C9H18O | |
MFCD00007030 | |
pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde | |
CHEBI:84268 | |
CCCCCCCCC=O |
Specifications
124-19-6 | |
79°C to 81°C (12 mmHg) | |
C9H18O | |
MFCD00007030 | |
pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde | |
GYHFUZHODSMOHU-UHFFFAOYSA-N | |
nonanal | |
31289 | |
142.24 | |
Nonanal |
0.825 | |
63°C (145°F) | |
1.424 | |
1236701 | |
Insoluble in water. | |
CCCCCCCCC=O | |
142.242 | |
CHEBI:84268 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 204-688-5
missing translation for 'rtecsNumber' : RA5700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only