Learn More
GPE Scientific™ Norell™ Thin-Wall Transparent Fluoropolymer FEP Tubing
Manufactured from special virgin grade thermoplastic FEP fluoropolymer
1520.00 SEK - 2380.00 SEK
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
13180833
|
GPE Scientific™
TWT-10 |
10.63 mm |
2380.00 SEK
Pack of 4 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
13120833
|
GPE Scientific™
TWT-4 |
4.91 mm |
1570.00 SEK
Pack of 4 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
13130833
|
GPE Scientific™
TWT-5 |
5.6 mm |
1620.00 SEK
Pack of 4 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
13140833
|
GPE Scientific™
TWT-6 |
6.55 mm |
1705.00 SEK
Pack of 4 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
13150833
|
GPE Scientific™
TWT-7 |
7.68 mm |
1810.00 SEK
Pack of 4 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
13160833
|
GPE Scientific™
TWT-8 |
8.54 mm |
1995.00 SEK
Pack of 4 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
13170833
|
GPE Scientific™
TWT-9 |
9.5 mm |
2195.00 SEK
Pack of 4 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
13110833
|
GPE Scientific™
TWT-3 |
3.5 mm |
1520.00 SEK
Pack of 4 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
Description
- Unlike PTFE tubing, this material can be heat sealed
- Heat sealing can offer extremely large possibilities for packaging and/or storage of samples (corrosives, liquids, solids, etc.)
- Fluoropolymer tubing can also be used where snug and tight fit over glass or metal tubing is required