missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norell™ Quartz Glass NMR Sample Tubes
For auto-sampling systems and high-throughput NMR
3420.00 SEK
Specifications
Product Type | NMR Sample Tube |
---|---|
Diameter (Metric) Outer | 4.97 mm |
Diameter (Metric) Inner | 4.2 mm |
Description
For auto-sampling systems and high-throughput NMR. The Norell Standard Series™ of NMR tubes has been developed for quality and precision. Virtually all bottom and wall variation has been eliminated, greatly improving shimmering times and sample reproducibility.Whether monitoring a reaction, scouting for a new compound, or doing routine NMR, the Standard Series™ is the tube of choice for today's scientist.All tubes have polished ends making them less susceptible to breaking. Coloured caps are included.Specifications
NMR Sample Tube | |
4.97 mm | |
4.2 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title