missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-PAGE™ SeeBlue™ Pre-stained Standard

Designed for use with pre-cast E-PAGE™ Electrophoresis System

Brand:  Invitrogen™ LC5700

Product Code. 10543843

  • 2470.00 SEK / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.


Includes: 500μL (50 applications of 10μL each) of the E-PAGE™ SeeBlue Pre-stained Standard is provided in loading buffer consisting of Tris-HCl, Formamide, and SDS.250μL (50 applications of 5μL each) of the E-PAGE™magicMark™ Unstained Protein Standard is provided in loading buffer consisting of 125mM Tris-HCl (pH 6.8), 10mM DTT, 17.4% Glycerol, 1% SDS, and 0.025% Bromophenol Blue
This item is not returnable. View return policy

Description

Description

  • Consists of three blue-dyed and two colored bands ranging in size from 21-261kDa and is suitable for monitoring gel and transfer runs
  • Consists of five recombinant proteins ranging in size from 20-220kDa, each with an IgG binding-site.
  • Can be detected with Coomassie™ stain on the gels or conjugated-antibody directly on western blots
  • Suitable for molecular weight estimation of unknown proteins
  • Gel Compatibility: E-PAGE

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contents: 500 μL (50 applications of 10 μL each) of the E-PAGE SeeBlue Pre-stained Standard

Storage buffer: Tris-HCl, formamide, and SDS

Storage: Stable for 4 months at +4°C

5
Yes
21 to 261 kDa
E-PAGE™ Gels
E-PAGE™, SeeBlue™
Wet Ice
Colorimetric
Protein Ladder
E-PAGE gels
3 colors: blue, orange, pink
261, 173, 97, 42, 21 kDa
500 μL
E-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.