missing translation for 'onlineSavingsMsg'
Learn More

NPM1 Mouse anti-Human, Clone: rNPM1/1901, Novus Biologicals™

Mouse Monoclonal Antibody

2480.00 SEK

Specifications

Antigen NPM1
Clone rNPM1/1901
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18324102
View Documents
Bio-Techne
NBP3-07449-20UG
20 μg
2480.00 SEK
20µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NPM1 Monoclonal specifically detects NPM1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin.
Specifications

Specifications

NPM1
0.2 mg/ml
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Mitotic Regulators, Phospho Specific, Signal Transduction, Tumor Suppressors
10 mM PBS with 0.05% BSA
4869
IgG1 κ
Protein A or G purified
rNPM1/1901
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
B23, MGC104254, NPMnucleophosmin/nucleoplasmin family, member 1, Nucleolar phosphoprotein B23, Nucleolar protein NO38, nucleophosmin, nucleophosmin (nucleolar phosphoprotein B23, numatrin), numatrin
Recombinant human NPM1 protein fragment (aa185-287) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.