missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin Mouse anti-Human, Clone: NCL/7338, Novus Biologicals™
Mouse Monoclonal Antibody
2470.00 SEK - 4910.00 SEK
Specifications
Antigen | Nucleolin |
---|---|
Clone | NCL/7338 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18337484
|
Bio-Techne
NBP3-13980-20UG |
20 μg |
2470.00 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18310106
|
Bio-Techne
NBP3-13980-100UG |
100 μg |
4910.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Nucleolin | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA Replication Transcription Translation and Splicing, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
Recombinant full-length human Nucleolin protein (Uniprot: P19338) | |
Primary | |
Store at 4°C. |
NCL/7338 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P19338 | |
4691 | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title