missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 96 PCR Clean-up Plates
Additional 96-well binding plates for proven, high-throughput clean-up of PCR products using the NucleoSpin™ 96 PCR Clean-up Kit or Core Kit.
Brand: Macherey-Nagel™ 740658.24S
This item is not returnable.
View return policy
Description
NucleoSpin™ 96 PCR Clean-up Kits
- Time-saving parallel clean-up of PCR products using a proven 96-well plate format
- Complete removal of primers and primer-dimers – optimal recovery
- Suitable for processing under vacuum and centrifugation
- Suitable for manual or automated use
- Innovative MN Wash Plate minimizes risk of cross-contamination
- NucleoSpin™ 96 PCR Clean-up Core Kits: basic content focused on automation platforms
Specifications
Room temperature (18 - 25 °C) | |
Silica Membrane Technology | |
Binding capacity: 15 μg | |
NucleoSpin 96 PCR Clean-up kit or core kit | |
96/Plate | |
24 Plates | |
45 min/plate |
96-well Plate | |
PCR Reaction Mixture | |
Elution volume: 75 - 150 μL | |
Clean-up of PCR products | |
Binding Plate | |
< 100 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction