missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Stool

Complete kits for quick and efficient isolation of total RNA from a variety of stool specimens using a combination of silica membrane technology and NucleoSpin™ Bead Tubes A (included). Kits available for 10 or 50 preps.

1435.00 SEK - 5385.00 SEK

Specifications

Format Mini Spin Column
Isolation Technology Silica Membrane Technology, NucleoSpin Bead Tubes
Elution Volume 100 μL
Sample Type Animal or Human Stool (Fresh or Frozen)
Efficiency Binding capacity: 200 μg
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
15815224
View Documents
Macherey-Nagel™
740130.10
10
1435.00 SEK
10 reactions
Please to purchase this item. Need a web account? Register with us today!
15875244
View Documents
Macherey-Nagel™
740130.50
50
5385.00 SEK
50 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RNA Stool kit enables efficient isolation of high quality RNA from any kind of stool sample. The kit combines NucleoZOL and a specific Lysis Buffer to chemically disrupt stool samples. The containing microbes are then disrupted with a mechanical lysis using the included Bead Tubes Type A.

  • Optimized workflow ensures a fast isolation of high quality RNA, including small RNA
  • Combination of mechanical homogenization and chemical lysis
  • Isolated RNA does not show any PCR inhibition due to Inhibitor Removal Columns
  • Suitable for various human and animal stool samples, fresh or frozen
Specifications

Specifications

Mini Spin Column
100 μL
Binding capacity: 200 μg
Isolation of RNA from stool
A260/A280: 1.9 - 2.1
RNA integrity number (RIN): > 7.5
10 - 30 μg
Silica Membrane Technology, NucleoSpin Bead Tubes
Animal or Human Stool (Fresh or Frozen)
High integrity RNA
Columns with Collection Tubes, NucleoZOL, Bead Tubes Type A, Inihibitor Removal Columns with Collection Tubes, Collection Tubes (2 mL), Collection Tubes (1.5 mL), rDNase, buffers
180 - 220 mg
70 min / 10 preps (preparation time)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • NucleoSpin RNA Stool (10)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : phenol, guanidium thiocyanate, rDNAse, 1,4-dioxane, ethanol

SDS
Documents

Documents

Product Certifications