missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoTraPCR kit for PCR clean up
Especially designed for direct purification of DNA fragments from PCR reactions without gel electrophoresis.
358.00 SEK - 1860.00 SEK
Specifications
Capacity | 0.6 μg/10 μL suspension |
---|---|
Final Product Type | CR Suspension |
Includes | Buffers |
Product Type | CR Suspension |
Sample Volume | <400 μL |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10008960
|
Macherey-Nagel™
740587 |
100 Preps |
1860.00 SEK
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10595571
|
Macherey-Nagel™
740587.10 |
10 Preps |
358.00 SEK
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NucleoTraP™CR extraction system is especially designed for direct purification of DNA fragments from PCR reactions without gel electrophoresis. DNA is reversibly adsorbed to the matrix with the aid of chaotropic agents. The NucleoTraP™CR extraction system can selectively bind DNA fragments ≥100bp, while the primer and primer-dimer molecules remain in solution. After two washing steps the DNA is eluted in low-salt elution buffer (5mM Tris/HCl, pH 8.5) or another low-salt medium (eg. water, pH 8.0). The eluted DNA shows a very high purity. All impurities such as enzymes, nucleotides and primer molecules are quantitatively removed. The eluted DNA can be used directly for all kinds of cloning experiments (ligation reactions, labelling reactions). DNA purified with NucleoTraP™CR gives excellent results in sequencing experiments using radioactive or fluorescent detection methods. Kit components: NucleoTraP™ suspension, buffers.- Silica matrix technology
- Yield: 70% to 80%
- Sample material: 20μL to 400μL PCR reaction mixture
- Elution volume: 20μL to 50μL
- Binding capacity: 6μg/10μL suspension
- Preparation time: 45 min./6 preps
- Fragment size: 100bp to 20kb
- Format: silica bead suspension
- No use of organic solvents For direct purification of PCR* products.
Specifications
0.6 μg/10 μL suspension | |
Buffers | |
<400 μL |
CR Suspension | |
CR Suspension |
Safety and Handling
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ NucleoTraPCR kit for PCR clean up