missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 10/11mL Polycarbonate Centrifuge Tubes
BIOPHARMA

Increase the effectiveness of centrifugation with these polycarbonate centrifuge tubes, which have been both RCF and tightness tested.

Brand:  Thermo Scientific™ 347708

5011.85 SEK valid until 2024-12-31
Use promo code "21693" to get your promotional price.


 View more versions of this product

Product Code. 10374701

  • 5600.00 SEK / Pack of 480
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Increase the effectiveness of centrifugation with Thermo Scientific™ Nunc™ 10/11mL Polycarbonate Centrifuge Tubes, which have been both RCF and tightness tested.

  • Made in a variety of formats for greater flexibility
  • Different bottom shapes to meet all needs
  • Include rim seal polypropylene screw caps
  • Fit most standard centrifuges
  • Relative centrifugal force tested (RCF)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

11 mL
480
Sterile
110 mm
Yes
Centrifuge Tube
Polycarbonate
80
Conical
Conical
16 mm
6000 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers