missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Phenylphenol, Na Salt,MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
CAS | 132-27-4 |
Molecular Formula | C12H9NaO |
Quantity | 100 g |
Synonym | sodium 1,1'-biphenyl-2-olate, sodium 2-biphenylate, sodium o-phenylphenate, 2-phenylphenol sodium salt, natriphene, orphenol, bactrol, o-phenylphenol sodium, sodium o-phenylphenol, dowicide a |
InChI Key | KSQXVLVXUFHGJQ-UHFFFAOYSA-M |
SMILES | C1=CC=C(C=C1)C2=CC=CC=C2[O-].[Na+] |
IUPAC Name | sodium;2-phenylphenolate |
Molecular Weight (g/mol) | 192.193 |
PubChem CID | 23675735 |
ChEBI | CHEBI:82371 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'