Learn More
RETSCH O-Ring for Test Sieves
Placed in the test sieve's recess to provide maximum stability and optimum sealing when used in sieve stacks. These o-rings for test sieves are available in several sizes, based upon the test sieve's diameter.
2870.00 SEK - 682710.00 SEK
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
11904442
|
RETSCH
05.114.0012 |
450 mm |
682710.00 SEK
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
11944432
|
RETSCH
05.114.0048 |
400 mm |
152790.00 SEK
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
10506232
|
RETSCH
05.114.0045 |
100 mm |
2870.00 SEK
Each |
Estimated Shipment: 20-09-2024
Log in to see stock. |
|||||
10009672
|
RETSCH
05.114.0047 |
305 mm |
3125.00 SEK
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
10581153
|
RETSCH
05.114.0077 |
200 mm |
10995.00 SEK
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
- Provides maximum stability and optimum sealing when used in sieve stacks
- Available in several sizes, based upon the test sieve's diameter
Specifications
O-Ring | |
1Each |