missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octan-1-ol, Pure, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Specifications
Specifications
Melting Point | -16°C |
Color | Colorless |
Boiling Point | 195°C |
Packaging | Plastic Coated Glass bottle |
Quantity | 2.5 L |
Vapor Density | 4.5 |
Formula Weight | 130.23g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- 1-Octanol
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- H412-Harmful to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'