Learn More
OHAUS™ Glass Fiber Pads for MB35, MB45, MB27, MB90 and MB120 Moisture Analyzers
Glass Fiber Pads for MB35, MB45, MB27, MB90 and MB120 Moisture Analyzers
Brand:  OHAUS™ 80850087
852.27 SEK valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 11894762
  • 1380.00 SEK / Pack of 200
Explore more special offers
Description
Description
  • Shape: round
  • Pack of 200
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MB35, MB45, MB27, MB90, MB120 Moisture Analyzer
Fiberglass
Pad
Special Offers
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Save up to 94% on Teaching Lab Essentials
    Promo Code25721
    Expires2025-05-01
    New Offer
    Save Now
Product Content Correction
Product Title
OHAUS™ Glass Fiber Pads for MB35, MB45, MB27, MB90 and MB120 Moisture Analyzers > Glass Fiber Pads for MB35, MB45, MB27, MB90 and MB120 Moisture Analyzers