Learn More
Kern Sohn OIML Class E2 Compact Weight Set, Plastic Case
Weight set constructed of polished stainless steel and housed in a plastic case.
Brand:  Kern Sohn 312-074
Encompass_Preferred
 View more versions of this product
Product Code. 17311883
  • 15854.06 SEK / 1 set
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Set of weights
  • Polished stainless steel
  • Compact
Specifications
Specifications
E2
Stainless Steel
Plastic
1 g to 2 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn OIML Class E2 Compact Weight Set, Plastic Case