Learn More
Kern Sohn OIML Class E2 Weight Set, Wooden Box
Weight set constructed of polished stainless steel and housed in a wooden box.
Brand:  Kern Sohn 313-06
Additional Details : Weight : 4.21000kg
 View more versions of this product
Product Code. 17321903
  • 20152.43 SEK / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
  • Set of weights
  • Polished stainless steel
  • Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic case
Specifications
Specifications
E2
Stainless Steel
Wood
1 mg to 1 kg