Learn More
Kern Sohn OIML Class F1 Weight Set, Wooden Box
Weight set constructed of polished stainless steel and housed in a wooden box.
Brand:  Kern Sohn 324-05
Encompass_Preferred
 View more versions of this product
Product Code. 17312063
  • 9907.33 SEK / 1 set
Estimated Shipment: 11-10-2024
to see stock.
Explore more special offers
Description
  • Knob shape
  • Polished stainless steel
  • Lined case
  • Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic box
Specifications
Specifications
F1
Stainless Steel
Wood
1 to 500 g