Learn More
Kern Sohn OIML Class F1 Weight Set, Wooden Box
Weight set constructed of polished stainless steel and housed in a wooden box.
Brand: Kern Sohn 324-03
This item is not returnable.
View return policy
Description
- Knob shape
- Polished stainless steel
- Lined case
- Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic box
Specifications
F1 | |
Stainless Steel | |
Wood | |
1 to 100 g |