Learn More
Kern Sohn OIML Class M1 ECO-Block Test Weight
Single test weight constructed of cast iron.
Brand: Kern Sohn T346-79-A
This item is not returnable.
View return policy
Description
- ECO rectangular shape
- Lacquered cast iron
Specifications
M1 | |
Cast Iron | |
50 kg | |
± 2500 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.