Learn More
Kern Sohn OIML Class M1 Weight Set, Wooden Box
Weight set constructed of finely turned stainless steel and housed in a wooden box.
Brand:  Kern Sohn 343-04
Encompass_Preferred
 View more versions of this product
Product Code. 17322193
  • 3321.21 SEK / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Description
  • Knob shape
  • Finely turned stainless steel
  • Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic box
Specifications
Specifications
M1
Stainless Steel
Wood
1 mg to 200 g
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.