Learn More
Ohaus™ OIML E1 Weight Set, NVLAP Certificate
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E1 Tolerances
19175.00 SEK - 158575.00 SEK
Specifications
Certifications/Compliance NVLAP Accredited Certificate
Includes Protective Case
View More Specs
Products 17
Product Code Brand Mass Price Quantity & Availability  
Product Code Brand Mass Price Quantity & Availability  
16612282
View Documents
Ohaus™
30390926
500 mg to 1 mg
19175.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16602282
View Documents
Ohaus™
30390924
50 g to 1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16692272
View Documents
Ohaus™
30390923
50 g to 1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16682272
View Documents
Ohaus™
30390922
100 g to 1 mg
44615.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16672272
View Documents
Ohaus™
30390921
100 g to 1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16612202
View Documents
Ohaus™
30390040
2 x 200 g to 1 mg
57870.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16602202
View Documents
Ohaus™
30390039
2 x 200 g to 1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16662272
View Documents
Ohaus™
30390918
500 g to 1 mg
62130.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16652272
View Documents
Ohaus™
30390917
500 g to 1 g
49115.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16642272
View Documents
Ohaus™
30390916
1 kg to 1 mg
73610.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16632272
View Documents
Ohaus™
30390915
1 kg to 1 g
55505.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16692192
View Documents
Ohaus™
30390038
2 x 2 kg to 1 mg
103430.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16682192
View Documents
Ohaus™
30390037
2 x 2 kg to 1 g
85205.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16622272
View Documents
Ohaus™
30390912
5 kg to 1 mg
131355.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16612272
View Documents
Ohaus™
30390911
5 kg to 1 g
110885.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16602272
View Documents
Ohaus™
30390909
10 kg to 1 mg
158575.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
16692262
View Documents
Ohaus™
30390908
10 kg to 1 g
144375.00 SEK
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
Description
Description
  • OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
  • Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
  • NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
Specifications
Specifications
NVLAP Accredited Certificate
Protective Case