Learn More
Ohaus™ OIML E2 Weight Set
Adjusted to Meet or Exceed OIML E2 Tolerances
Brand:  Ohaus™ 30518036
8851.87 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16662372
  • 8900.00 SEK / Each
Estimated Shipment: 05-05-2025
to see stock.
Explore more special offers
Description
  • Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Certificate
50 g to 1 g
8/Pk.
Troemner Alloy 8
Protective Case
Documents
One moment while we fetch your results.
Product Title
Ohaus™ OIML E2 Weight Set > 50 g - 1 g