missing translation for 'onlineSavingsMsg'
Learn More

Oleic acid, 99%, MP Biomedicals™
BIOPHARMA

800.00 SEK - 4335.00 SEK

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (9E)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11462572
BIOPHARMA
View Documents
MP Biomedicals
0215178101
1 g
800.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
11472572
BIOPHARMA
View Documents
MP Biomedicals
0215178105
5 g
1435.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
11482572
BIOPHARMA
View Documents
MP Biomedicals
0215178125
25 g
4335.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

112-80-1
282.47
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
445639
(9E)-octadec-9-enoic acid
C18H34O2
MFCD00064242
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
CHEBI:16196
CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

112-80-1
0.89g/L
MFCD00064242
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
(9E)-octadec-9-enoic acid
445639
282.52
Solid
White
C18H34O2
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
CCCCCCCC\C=C\CCCCCCCC(O)=O
282.47
CHEBI:16196
0.99
Oleic acid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.