missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oligomycin, >98%, For TLC analysis, MP Biomedicals™
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Specifications
Specifications
Type | Oligomycin |
Physical Form | Powder |
Formula Weight | ≈789.3 |
Percent Purity | >98% |
Color | White |
Quantity | 1 mg |
Chemical Name or Material | Oligomycin, For TLC analysis |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'