Learn More
B Braun™ Ominifix™ Solo Single-use Irrigation Syringe
Manufactured from polypropylene. Omnifix™ Solo Single-use Irrigation Syringe with highly transparent barrel and high contrast black graduation provides ideal readability. Plunger stopper includes double sealing ring for slow aspiration and injection.
Brand: B Braun™ 4616308F
This item is not returnable.
View return policy
Description
- Highly transparent barrel
- High contrast black graduation
- Permanent marking Ideal readability
- Safe plunger backstop
- Plunger stopper with double sealing ring for slow aspiration and injection, qualified according to ISO 10993 resp. DIN EN 30993 Qualified according to ISO 7886-1 olypropylene Latex-free PVC-free
Specifications
EN ISO 7886-1 | |
Eccentric, Luer-slip | |
1/Ea. | |
Sterile |
Polypropylene | |
Single-use | |
30 mL |