missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jaytec Glass™ Open Top Borosilicate Glass Dropping Funnels
Open Top Borosilicate Glass Dropping Funnels for easy refilling in situ. The stopcock permits carefully controlled flow of reagents.
Brand: Jaytec Glass™ FTP 050
This item is not returnable.
View return policy
Description
- Open top
- Cylindrical shape
- With interchangeable PTFE stopcock
Specifications
30 mm | |
8 mm | |
232 mm | |
PTFE stopcock key | |
Dropping funnel |
50 mL | |
Borosilicate Glass | |
No | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction