Learn More
Jaytec Glass™ Open Top Borosilicate Glass Dropping Funnels
Open Top Borosilicate Glass Dropping Funnels for easy refilling in situ. The stopcock permits carefully controlled flow of reagents.
Brand:  Jaytec Glass™ FTP250
 View more versions of this product
Product Code. 12358577
  • 544.00 SEK / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Open top
  • Cylindrical shape
  • With interchangeable PTFE stopcock
Specifications
Specifications
52 mm
10 mm
342 mm
PTFE stopcock key
Dropping funnel
250 mL
Borosilicate Glass
No
Yes