Learn More
Hettich™ Optional Lid for Rotors
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 2423
Additional Details : Weight : 0.80000kg
Description
- Lid with bio-containment
- For rotors no. 1154-L, 1420-B, 2219-A, 2424-B, 2426-B, 2427-A and 2430-B
- Autoclavable
- Phenol-resistant
Specifications
Biocontainment Lid | |
Hettich Rotors | |
No | |
Autoclavable |