missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR51S1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1925.00 SEK - 4460.00 SEK
Specifications
Antigen | OR51S1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18321484
|
Bio-Techne
NBP3-09864-25UL |
25 μg |
1925.00 SEK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18318786
|
Bio-Techne
NBP3-09864-100UL |
100 μg |
4460.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OR51S1 Polyclonal specifically detects OR51S1 in Human samples. It is validated for Western Blot.Specifications
OR51S1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
olfactory receptor 51S1, Olfactory receptor OR11-24, olfactory receptor, family 51, subfamily S, member 1, OR11-24 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR51S1 (NP_001004758). Peptide sequence LDPLLIFFSYGLIGKVLQGVESREDRWKAGQTCAAHLSAVLLFYIPMILL | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
119692 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title