Learn More
BD Oral Syringes
Easy to read scale markings in mL
1690.00 SEK - 3775.00 SEK
Product Code | Brand | Color | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume (Metric) | Price | Quantity & Availability | ||||
15246876
|
BD
305209 |
Amber | 10 mL |
3775.00 SEK
Pack of 500 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15296866
|
BD
305207 |
Amber | 1 mL |
3000.00 SEK
Pack of 500 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13498479
|
BD
305210 |
Amber | 3 mL |
2580.00 SEK
Pack of 500 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13107653
|
BD
305208 |
Amber | 5 mL |
1690.00 SEK
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Oral Syringes
Specifications
Yes | |
Oral Syringe | |
Non-sterile |