Learn More
Org-26576, MedChemExpress
MedChemExpress Org-26576 is a AMPA receptor positive allosteric modulator.
Brand: Medchem Express HY-101216
This item is not returnable.
View return policy
Description
Org-26576 is a AMPA receptor positive allosteric modulator.Specifications
Org-26576 | |
Off-White | |
C11H12N2O2 | |
DMSO : 25 mg/mL (122.41 mM; Need ultrasonic) | |
204.23 | |
98.58% | |
Solid |
1026791-61-6 | |
Research | |
100 mg | |
O=C1N2[C@](CCC2)([H])COC3=NC=CC=C13 | |
204.23 | |
Research | |
Neuroscience-Neuromodulation |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Org-26576, MedChemExpress