Learn More
Thermo Scientific™ Orion™ Calibration Sleeves for Dissolved Oxygen Probes
Calibrate and store probes with Thermo Scientific™ Orion™ Calibration Sleeves for Dissolved Oxygen Probes.
Brand: Thermo Scientific™ 086021A
Additional Details : Weight : 0.01000kg
Description
Calibrate and store probes with Thermo Scientific™ Orion™ Calibration Sleeves for Dissolved Oxygen Probes.
Provide optimum stable environment for calibration.Specifications
Calibration Sleeve with Stand | |
Dissolved Oxygen Probe | |
Calibration Sleeve |