missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ortho-Phosphoric Acid, 85%, ACS Reagent Grade, Scharlab
Click to view available options
Quantity:
1 L
2.5 L
Description
- Synonyms: Orthophosphoric acid
- Molecular Formula: H3PO4
- Molar Mass: 98.00g/mol
- CAS: 7664-38-2
- EIN ECS No.: 231-633-2
- Passes test for substances precipitated with ammonia
Specifications
Specifications
Appearance of Solution | Clear and colorless |
Melting Point | ∼21°C |
Boiling Point | ∼158°C |
Density | 1.71g/cm3 |
Identification | Passes Test |
Vapor Pressure | 2.2 hPa (at 25°C) |
Quantity | 1 L |
Physical Form | Liquid |
Solubility Information | Miscible in water (20°C) |
Formula Weight | 98.00g/mol |
Show More |
Safety and Handling

Product Identifier
- ortho-Phosphoric acid, 85%, reagent grade, ACS, ISO, Reag. Ph Eur
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Phosphoric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ortho-Phosphoric Acid, 85%, ACS Reagent Grade, Scharlab >
Spot an opportunity for improvement?Share a Content Correction