missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otx1 Rabbit anti-Human, Mouse, Rat, Clone: 6H7J9, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | Otx1 |
---|---|
Clone | 6H7J9 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18354745
|
Bio-Techne
NBP3-15884-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18323293
|
Bio-Techne
NBP3-15884-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Otx1 Monoclonal antibody specifically detects Otx1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Otx1 | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ38361, homeobox protein OTX1, MGC15736, orthodenticle (Drosophila) homolog 1, orthodenticle homeobox 1, Orthodenticle homolog 1, orthodenticle homolog 1 (Drosophila) | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Otx1 (P32242). MMSYLKQPPYGMNGLGLAGPAMDLLHPSVGYPATPRKQRRERTTFTRSQLDVLEALFAKTRYPDIFMREEVALKINLPESRVQVWFKNRRAKCRQQQQSG | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
6H7J9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5013 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title