missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic acid solution, Volumetric, 0.5 M (COOH)2 (1N), Honeywell Fluka™
Volumetric, 0.5 M (COOH)2 (1N)
1702.40 SEK - 6752.17 SEK
Chemical Identifiers
CAS | 144-62-7 |
---|---|
Molecular Formula | C2H2O4 |
Molecular Weight (g/mol) | 90.034 |
InChI Key | MUBZPKHOEPUJKR-UHFFFAOYSA-N |
Synonym | ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico |
PubChem CID | 971 |
ChEBI | CHEBI:16995 |
IUPAC Name | oxalic acid |
SMILES | C(=O)(C(=O)O)O |
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
144-62-7 | |
90.034 | |
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico | |
CHEBI:16995 | |
C(=O)(C(=O)O)O |
C2H2O4 | |
MUBZPKHOEPUJKR-UHFFFAOYSA-N | |
971 | |
oxalic acid |
Specifications
144-62-7 | |
HOOCCOOH | |
NONH for all modes of transport | |
MUBZPKHOEPUJKR-UHFFFAOYSA-N | |
oxalic acid | |
971 | |
90.03g/mol |
C2H2O4 | |
MFCD00002573 | |
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico | |
C(=O)(C(=O)O)O | |
90.034 | |
CHEBI:16995 | |
Oxalic acid solution |
Safety and Handling
P280-P305 + P351 + P338