Learn More
Thermo Scientific™ Oxoid™ Legionella MWY Selective Supplement, for 100mL medium
Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.
Brand: Thermo Scientific™ SR0118E
This item is not returnable.
View return policy
Description
Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.
Add to Legionella CYE Agar Base, Part No. CM0655B