Learn More
Thermo Scientific™ Oxoid™ Ringers Solution Tablets
Oxoid Ringers Solution Tablets is used for the preparation of isotonic diluents for bacterial cells and bacteriological specimens.
Brand: Thermo Scientific™ BR0052G
377.37 SEK valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Additional Details : Weight : 0.14400kg
Description
Oxoid Ringers Solution Tablets is used for the preparation of isotonic diluents for bacterial cells and bacteriological specimens.
Each tablet prepares 500mL solution.Specifications
Industrial Reference; AFNOR, BSI, IDF, ISO, NMKL. | |
Tablet | |
Each |
Ringers Solution Tablets | |
Media Supplement | |
Each tab for 500mL medium |
For Laboratory Use Only