missing translation for 'onlineSavingsMsg'
Learn More

Oxolinic acid, 98%, Thermo Scientific™

Product Code. 15428079
Click to view available options
Quantity:
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

Targets gram-negative bacteria

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 14698-29-4
Molecular Formula C13H11NO5
MDL Number MFCD00056775
Beilstein 620635
Merck Index 14,6965
Synonym 5,8-Dihydro-5-ethyl-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid
Solubility Information Soluble in 0.5N NaOH with warming
InChI Key KYGZCKSPAKDVKC-UHFFFAOYSA-N
SMILES CCN1C=C(C(O)=O)C(=O)C2=CC3=C(OCO3)C=C12
IUPAC Name 5-ethyl-8-oxo-2H,5H,8H-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid
Molecular Weight (g/mol) 261.23
PubChem CID 4628
ChEBI CHEBI:138856
Formula Weight 261.23
Percent Purity 98%
Color White
Quantity 5g
Flash Point >110°C (230°F)
Sensitivity Light sensitive
Boiling Point 473°C
Chemical Name or Material Oxolinic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'