missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl 2-acetamido-2-deoxy-a-D-glucopyranoside, MP Biomedicals

Product Code. 11429191
Click to view available options
Quantity:
10 mg
25 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10139-02-3
Molecular Formula C14H18N2O8
Molecular Weight (g/mol) 342.304
InChI Key OMRLTNCLYHKQCK-KSTCHIGDSA-N
Synonym 4'-nitrophenyl-2-acetamido-2-deoxy-alpha-d-glucopyranoside, p-nitrophenyl n-acetyl-alpha-d-glucosaminide, n-2r,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-a-d-glucopyranoside, 4-nitrophenyl 2-acetamido-2-deoxy-alpha-d-glucopyranoside, n-2r,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-, a-d-glucopyranoside, 4-nitrophenyl n-acetyl-, a-d-glucosaminide, p-nitrophenyl-n-acetyl-alpha-d-glucosaminide, p-nitrophenyl 2-acetamido-2-deoxy-
PubChem CID 82398
IUPAC Name N-[(2R,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1OC2=CC=C(C=C2)[N+](=O)[O-])CO)O)O

Specifications

Quantity 10 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.