missing translation for 'onlineSavingsMsg'
Learn More

p57Kip2Ab-6 Mouse Monoclonal Antibody, Epredia™

Deliver accurate, reproducible results with the Epredia™ p57Kip2 Ab-6, Mouse Monoclonal Antibody.

1815.00 SEK - 10120.00 SEK

Specifications

Antigen p57superscriptKip2superscript Ab-6
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
Host Species Mouse
View More Specs

Products 7
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12668776
View Documents Promotion Details
Epredia
MS-1062-P0
Concentrated 100 μL
1815.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
12688776
View Documents
Epredia
MS-1062-P1ABX
Concentrated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12698776
View Documents
Epredia
MS-1062-PABX
Concentrated 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12678776
View Documents Promotion Details
Epredia
MS-1062-P1
Concentrated 500 μL
5070.00 SEK
500µL
Please to purchase this item. Need a web account? Register with us today!
12658776
View Documents Promotion Details
Epredia
MS-1062-P
Concentrated 1 mL
10120.00 SEK
1mL
Please to purchase this item. Need a web account? Register with us today!
12628786
View Documents Promotion Details
Epredia
MS-1062-R7
Prediluted 7 mL
5135.00 SEK
7mL
Please to purchase this item. Need a web account? Register with us today!
12638786
View Documents
Epredia
MS-1062-RQ
Ready-to-Use 12 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

p57Kip2 (or CDKN1C) is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. The gene encoding human p57Kip2 is located on chromosome 11p15.5, a region implicated in both sporadic cancers, Wilm's tumor, and Beckwith-Wiedemann syndrome (BWS), a cancer syndrome, making it a tumor suppressor candidate. BWS is characterized by numerous growth abnormalities and an increased risk of childhood tumors. Several types of childhood tumors including Wilms' tumor, adrenocortical carcinoma and rhabdomyosarcoma display a specific loss of maternal 11p15 alleles, suggesting that genomic imprinting plays an important part. This region also contains two other imprinted genes, insulin-like growth factor II (IGF-II) and H19, both of which seem to be implicated in adrenal neoplasms.

Host Species: Mouse

Clone: 57P06; same as KP10

Isotype: IgG2b/κ

Species Reactivity: Human and Mouse. Others not known.

Epitope: Not determined

Immunogen: Recombinant human p57Kip2 protein

Molecular Weight: 57kDa

Positive Control: LS174T cells, Colon CA or placenta

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunoprecipitation (Native verified)

Specifications

Specifications

p57superscriptKip2superscript Ab-6
Monoclonal
Mouse
Cell Cycle
Primary
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
IVD
Recombinant human p57superscriptKip2superscript protein
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.