missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancuronium bromide, MP Biomedicals™
Click to view available options
Quantity:
10 mg
50 mg
100 mg
0.25 g
Specifications
Specifications
Color | White |
Quantity | 10 mg |
Formula Weight | 732.7 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Pancuronium bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'