Learn More
PANX1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12595-1-AP-150UL
 View more versions of this product
Product Code. 16880384
  • 4721.67 SEK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The pannexin gene family encodes a second class of putative gap junction proteins and are highly conserved in invertebrates and mammals. Pannexins (Panx) are four-pass transmembrane proteins that oligomerize to form large pore ion and metabolite-permeable channels. Pannexin-1 (PANX1) and Pannexin-3 are closely related, while Pannexin-2 is a more distant relation. PANX1 is a transmembrane protein that forms a mechanosensitive ATP-permeable channel between adjacent cells and in the endoplasmic reticulum. PANX1 may play a role as a Ca2+ -leak channel to regulate ER Ca2+ homeostasis and regulates neural stem and progenitor cell proliferation.
Specifications
Specifications
PANX1
Polyclonal
Unconjugated
PANX1
MRS1, pannexin 1, PANX1, PX1, UNQ2529
Rabbit
Antigen Affinity Chromatography
RUO
24145
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96RD7
PANX1
PANX1 Fusion Protein Ag3174
150 μL