missing translation for 'onlineSavingsMsg'
Learn More

Paraffin, Pure, Soft Yellow, SLR, Fisher Chemical

Product Code. 10427650
Click to view available options
Quantity:
500 g
Packaging:
Plastic container
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 64771-71-7
Melting Point 38°C
Color White to Yellow
Boiling Point 200°C
Packaging Plastic container
Molecular Formula C10H21
MDL Number 131611
Quantity 500 g
Molecular Weight (g/mol) 141.28
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'