missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513311
3000.15 SEK valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Poly(ADP-ribosyl)ation is an immediate DNA-damage-dependent post-translational modification of histones and other nuclear proteins that contributes to the survival of injured proliferating cells. Poly(ADP-ribose) polymerases (PARPs) now constitute a large family of 18 proteins, encoded by different genes and displaying a conserved catalytic domain in which PARP-1 (113 kDa), the founding member, and PARP-2 (62 kDa) are so far the sole enzymes whose catalytic activity has been shown to be immediately stimulated by DNA strand breaks. A large repertoire of sequences encoding novel PARPs now extends considerably the field of poly(ADP-ribosyl)ation reactions to various aspects of the cell biology including cell proliferation and cell death. Some of these new members interact with each other, share common partners and common subcellular localizations suggesting possible fine tuning in the regulation of this post-translational modification of proteins.
Specifications
PARP12 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8BZ20, Q9H0J9 | |
PARP12 | |
KLH conjugated synthetic peptide between 454-483 amino acids from the C-terminal region of human Parp12 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PARP12 | |
9930021O16; AA409132; AA536654; ADP-ribosyltransferase diphtheria toxin-like 12; ARTD12; MST109; MSTP109; PARP12; PARP-12; poly (ADP-ribose) polymerase family, member 12; poly [ADP-ribose] polymerase 12; poly ADP-ribose polymerase 12; poly(ADP-ribose) polymerase family member 12; Protein mono-ADP-ribosyltransferase PARP12; ZC3H1; Zc3hdc1; Zinc finger CCCH domain-containing protein 1; zinc finger CCCH type domain containing 1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
243771, 64761 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PARP12 Polyclonal Antibody, Invitrogen™ > 400 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction