Learn More
Erlab™ Particulate Filter
Erlab™ Particulate Filter can be use with a range of filtrating fume hoods and storage cabinets.
Brand:  Erlab™ I1019000001
Additional Details : Weight : 5.00000kg
Product Code. 15991360
  • 1176.60 SEK / Each
Estimated Shipment: 12-08-2024
to see stock.
Description
Description
  • Filter HP - HEPA H14 : This filtration technology traps particles larger than 0.3 μm to optimize HEPA H14 filter life performance
  • For use with powders
Specifications
Specifications
HEPA H14
CaptairFlex XLS 633