Learn More
Poulten Graf™ Pasteur Pipets
Conforms to BS 5732
Brand:  Poulten Graf™ D814
Product Code. 10488791
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Nominal rate of 30 drops/mL to 35 drops/mL. BS 5732.
Specifications
Specifications
Glass
270 mm
Single-use
1000 Pack
Pasteur Pipet