Learn More
Poulten Graf™ Pasteur Pipets
Conforms to BS 5732
Brand: Poulten Graf™ D814
This item is not returnable.
View return policy
Description
Nominal rate of 30 drops/mL to 35 drops/mL. BS 5732.Specifications
Glass | |
270 mm | |
Single-use |
1000 Pack | |
Pasteur Pipet |